L11125.09
L11125.18
L11125.09EA
48.9
EUR
InStock
L11125.09
L11125.18
Methyl-2-amino-4-chlorbenzoat 98%
Methyl-2-amino-4-chlorbenzoat
Purity: 98%
CAS No.: 5900-58-3
Molecular Formula: C8H8ClNO2
Formula Weight: 185.61
Melting Point: 66-68°
Beilstein Reference: 1072895