L11457.14
L11457.22
L11457.14EA
59.7
EUR
InStock
L11457.14
L11457.22
4-Hydroxy-6-methyl-2H-pyran-2-on 98%
4-Hydroxy-6-methyl-2-pyrone
Purity: 98%
CAS No.: 675-10-5
Molecular Formula: C6H6O3
Formula Weight: 126.11
Melting Point: ca 186° dec.
Beilstein Reference: 113815
MDL No.: MFCD00006641