L11646.06
L11646.14
L11646.03
L11646.06EA
52.2
EUR
InStock
L11646.06
L11646.14
L11646.03
3-(Trifluormethyl)phenylisothiocyanat ≥98%
3-(Trifluormethyl)phenylisothiocyanat
Purity: 98%
CAS No.: 1840-19-3
Molecular Formula: C8H4F3NS
Formula Weight: 203.19
Flash Point: 95°(203°F)
Refractive Index: 1.5547
Odour: Pungent
Boiling Point: 208°
Beilstein Reference: 781205
UN No.: 2922
MDL No.: MFCD00004806