L11753.09
L11753.18
L11753.09EA
77.2
EUR
InStock
L11753.09
L11753.18
1-Chlor-3,3-dimethylbutan ≥97%
1-Chloro-3,3-dimethylbutane
Purity: 97%
CAS No.: 2855-08-5
Molecular Formula: C6H13Cl
Formula Weight: 120.62
Flash Point: 7°(44°F)
Refractive Index: 1.4170
Boiling Point: 116-118°
Beilstein Reference: 1731465
UN No.: 1993
MDL No.: MFCD00018980