L11842.22
L11842.36
L11842.22EA
35.9
EUR
InStock
L11842.22
L11842.36
EDTA Dikaliumsalz Dihydrat 99%
EDTA Dikaliumsalz Dihydrat
Purity: 99%
CAS No.: 25102-12-9
Molecular Formula: C10H14K2N2O8.2H2O
Formula Weight: 404.46 (368.43anhy)
Odour: Odourless
Melting Point: ca 272° dec.
MDL No.: MFCD00150036