L11965.14
L11965.22
L11965.14EA
39
EUR
InStock
L11965.14
L11965.22
Sodium-4-aminoazobenzene-4'-sulphonate 90%, tech.
Sodium-4-aminoazobenzene-4'-sulphonate
Purity: 90%
CAS No.: 2491-71-6
Molecular Formula: C12H10N3NaO3S
Formula Weight: 299.29
Beilstein Reference: 8098548
MDL No.: MFCD00035564