L12075.09
L12075.09EA
78
EUR
InStock
L12075.09
Propargyl-p-toluolsulfonat ≥97%
Propargyl-p-toluolsulfonat
Purity: 97%
CAS No.: 6165-76-0
Molecular Formula: C10H10O3S
Formula Weight: 210.25
Refractive Index: 1.5270
Boiling Point: 117-121°/0.3mm
MDL No.: MFCD00078365