L12147.09
L12147.09EA
43.3
EUR
InStock
L12147.09
3,5-Dichlor-2-hydroxybenzolsulfonylchlorid ≥97%
3,5-Dichlor-2-hydroxybenzolsulfonylchlorid
Purity: 97%
CAS No.: 23378-88-3
Molecular Formula: C6H3Cl3O3S
Formula Weight: 261.51
Melting Point: 78-80°
Beilstein Reference: 2940993
UN No.: 3261
MDL No.: MFCD00007432