L12270.06
L12270.14
L12270.06EA
69.8
EUR
InStock
L12270.06
L12270.14
Malonic acid dihydrazide 99%
Malonic acid dihydrazide
Purity: 99%
CAS No.: 3815-86-9
Molecular Formula: C3H8N4O2
Formula Weight: 132.12
Melting Point: 152-156°
Beilstein Reference: 1072337
MDL No.: MFCD00041268