L12355.03
L12355.06
L12355.03EA
35
EUR
InStock
L12355.03
L12355.06
1-(4-(Trifluormethyl)phenyl)thioharnstoff ≥98%
1-(4-(Trifluormethyl)phenyl)thioharnstoff
Purity: 98%
CAS No.: 1736-72-7
Molecular Formula: C8H7F3N2S
Formula Weight: 220.22
Odour: Odourless
Melting Point: 142-146°
Beilstein Reference: 2695479
UN No.: 2811
MDL No.: MFCD00041189