L12576.03
L12576.06
L12576.03EA
34.2
EUR
InStock
L12576.03
L12576.06
α,α,α-Trifluor-p-tolylisocyanat ≥98%
α,α,α-Trifluor-p-tolylisocyanat
Purity: 98%
CAS No.: 1548-13-6
Molecular Formula: C8H4F3NO
Formula Weight: 187.12
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Refractive Index: 1.4710
Odour: Pungent
Boiling Point: 91-94°/35mm
Beilstein Reference: 881041
UN No.: 2285
MDL No.: MFCD00002028