L13328.03
L13328.06
L13328.03EA
193
EUR
InStock
L13328.03
L13328.06
4,4'-Biphenyldiboronsäure 95%
4,4'-Biphenyldiboronsäure
Purity: 95%
CAS No.: 4151-80-8
Molecular Formula: C12H12B2O4
Formula Weight: 241.85
Melting Point: ca 300° dec.
Beilstein Reference: 2941876