L13461.14
L13461.14EA
185
EUR
InStock
L13461.14
3,4,5-Trimethoxybenzylamin 96%
3,4,5-Trimethoxybenzylamin
Purity: 96%
CAS No.: 18638-99-8
Molecular Formula: C10H15NO3
Formula Weight: 197.24
Flash Point: 110°(230°F)
Refractive Index: 1.5480
Boiling Point: 120-122°/0.05m
Beilstein Reference: 1107570
UN No.: 2735
MDL No.: MFCD00017150