L13597.MD
L13597.MDEA
37.5
EUR
InStock
L13597.MD
Pentifyllin ≥98%
Pentifyllin
Purity: 98+%
CAS No.: 1028-33-7
Molecular Formula: C13H20N4O2
Formula Weight: 264.33
Melting Point: 80-81°
Beilstein Reference: 270632
Merck Reference: 14,7127
MDL No.: MFCD00041424