L13756.03
L13756.06
L13756.14
L13756.03EA
42.6
EUR
InStock
L13756.03
L13756.06
L13756.14
Pyrrol-2-carbonsäure ≥99%
Pyrrol-2-carbonsäure
Purity: 99%
CAS No.: 634-97-9
Molecular Formula: C5H5NO2
Formula Weight: 111.10
Melting Point: ca 205° subl.
Beilstein Reference: 80825