L13987.03
L13987.MD
L13987.03EA
73.5
EUR
InStock
L13987.03
L13987.MD
(+)-Menthyl acetate 99%
(+)-Menthyl acetate
Purity: 99%
CAS No.: 5157-89-1
Molecular Formula: C12H22O2
Formula Weight: 198.31
Flash Point: 77°(170°F)
Refractive Index: 1.4470
Boiling Point: 229-230°
Beilstein Reference: 1909419
Optical Rotation: +80.5° (c=8 in benzene)
MDL No.: MFCD00062977