L14049.03
L14049.06
L14049.03EA
88.4
EUR
InStock
L14049.03
L14049.06
4-Carbethoxyhexafluorobutyryl chloride 97%
4-Carbethoxyhexafluorobutyryl chloride
Purity: 97%
CAS No.: 18381-53-8
Molecular Formula: C7H5ClF6O3
Formula Weight: 286.56
Refractive Index: 1.3590
Boiling Point: 158-160°
Beilstein Reference: 1884844
UN No.: UN3265
MDL No.: MFCD00054671