L14059.03
L14059.06
L14059.MD
L14059.03EA
58
EUR
InStock
L14059.03
L14059.06
L14059.MD
(1R,2R)-(-)-1,2-Diaminocyclohexan 98%
(1R,2R)-(-)-1,2-Diaminocyclohexan
Purity: 98%
CAS No.: 20439-47-8
Molecular Formula: C6H14N2
Formula Weight: 114.19
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Odour: Amine-like
Melting Point: 40-43°
Boiling Point: 104-110°/40mm
Beilstein Reference: 4780911
Optical Rotation: -25.5° (c=5 in 1N HCl)
UN No.: 3259
MDL No.: MFCD00062985