L14151.03
L14151.06
L14151.14
L14151.03EA
38.1
EUR
InStock
L14151.03
L14151.06
L14151.14
(R)-(-)-Methyl-3-hydroxybutyrat ≥98%
(R)-(-)-Methyl-3-hydroxybutyrat
Purity: 98%
CAS No.: 3976-69-0
Molecular Formula: C5H10O3
Formula Weight: 118.13
Flash Point: 73°(163°F)
Refractive Index: 1.4220
Odour: Pleasant
Boiling Point: 56-58°/11mm
Beilstein Reference: 3648161
Optical Rotation: -48.5° (c=1.3 in chloroform)
MDL No.: MFCD00063289