L14224.22
L14224.14
L14224.22EA
207
EUR
InStock
L14224.22
L14224.14
Diethoxymethylvinylsilan ≥97%
Diethoxymethylvinylsilan
Purity: 97%
CAS No.: 5507-44-8
Molecular Formula: C7H16O2Si
Formula Weight: 160.29
Flash Point: 17°(62°F)
Refractive Index: 1.400
Boiling Point: 133-134°
Beilstein Reference: 970124
UN No.: 1993
MDL No.: MFCD00009067