L14324.03
L14324.MD
L14324.03EA
74.8
EUR
InStock
L14324.03
L14324.MD
1-Fluorpyridiniumtrifluormethansulfonat 95%
1-Fluoropyridiniumtrifluoromethanesulphonate
Purity: 95%
CAS No.: 107263-95-6
Molecular Formula: C6H5F4NO3S
Formula Weight: 247.17
Melting Point: 180-185°
Beilstein Reference: 4222821
UN No.: 3261
MDL No.: MFCD00013458