L14342.06
L14342.14
L14342.22
L14342.06EA
84.9
EUR
InStock
L14342.06
L14342.14
L14342.22
Chrom(II)chlorid, wasserfrei 97%
Chrom(II)chlorid
Purity: 97%
CAS No.: 10049-05-5
Molecular Formula: CrCl2
Formula Weight: 122.90
Odour: Odourless
Melting Point: 824°
UN No.: 3260
MDL No.: MFCD00010947