L14583.AC
L14583.AA
L14583.ACEA
250
EUR
InStock
L14583.AC
L14583.AA
(S)-(-)-2-Methyl-CBS-oxazaborolidin 1 M in Toluol
(S)-(-)-2-Methyl-CBS-oxazaborolidin
CAS No.: 112022-81-8
Molecular Formula: C18H20BNO
Formula Weight: 277.17
Flash Point: 4°(39°F)
UN No.: 1993
MDL No.: MFCD00078439