L14610.03
L14610.06
L14610.03EA
46.9
EUR
InStock
L14610.03
L14610.06
6-Methoxy-2-naphthoesäure ≥98%
6-Methoxy-2-naphthoesäure
Purity: 98+%
CAS No.: 2471-70-7
Molecular Formula: C12H10O3
Formula Weight: 202.21
Melting Point: 192-194°
Beilstein Reference: 2211233