L15530.03
L15530.MD
L15530.03EA
119
EUR
InStock
L15530.03
L15530.MD
3-Isopropylphenylboronsäure ≥99%
3-Isopropylphenylboronsäure
Purity: 99%
CAS No.: 216019-28-2
Molecular Formula: C9H13BO2
Formula Weight: 164.01
Flash Point: 110°(230°F)
Melting Point: ca 75-80°
Beilstein Reference: 3090001
MDL No.: MFCD01074634