L15825.14
L15825.22
L15825.14EA
17.6
EUR
InStock
L15825.14
L15825.22
2-Hydroxy-m-toluylsäure ≥99%
2-Hydroxy-m-toluylsäure
Purity: 99%
CAS No.: 83-40-9
Molecular Formula: C8H8O3
Formula Weight: 152.15
Odour: Odourless
Melting Point: 163-167°
Beilstein Reference: 2086811
Merck Reference: 14,2581
MDL No.: MFCD00002448