L16015.AE
L16015.AP
L16015.0F
L16015.AEEA
24.1
EUR
InStock
L16015.AE
L16015.AP
L16015.0F
2,6-Dimethyl-4-heptanon ≥90%
2,6-Dimethyl-4-heptanon
Purity: 90% (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)
CAS No.: 108-83-8
Molecular Formula: C9H18O
Formula Weight: 142.24
Flash Point: 47°(117°F)
Refractive Index: 1.4120
Odour: Unpleasant
Melting Point: -42°
Boiling Point: 163-173°
Beilstein Reference: 1743163
UN No.: 1224
MDL No.: MFCD00008940