L16313.03
L16313.06
L16313.03EA
85.6
EUR
InStock
L16313.03
L16313.06
N-(4-Chloro-2-butynyl)phthalimide 97%
N-(4-Chloro-2-butynyl)phthalimide
Purity: 97%
CAS No.: 4819-69-6
Molecular Formula: C12H8ClNO2
Formula Weight: 233.66
Odour: Nearly Odourless
Melting Point: 115-120°
Beilstein Reference: 179626
UN No.: UN3261
MDL No.: MFCD01318120