L16354.06
L16354.14
L16354.06EA
59.5
EUR
InStock
L16354.06
L16354.14
3-Bromphenylboronsäure ≥98%
3-Bromphenylboronsäure
Purity: 98+%
CAS No.: 89598-96-9
Molecular Formula: C6H6BBrO2
Formula Weight: 200.83
Melting Point: 164-168°
Beilstein Reference: 3127104