L17008.03
L17008.06
L17008.03EA
48.9
EUR
InStock
L17008.03
L17008.06
3-(4-Fluorphenyl)propionsäure ≥97%
3-(4-Fluorphenyl)propionsäure
Purity: 97%
CAS No.: 459-31-4
Molecular Formula: C9H9FO2
Formula Weight: 168.17
Melting Point: 88-92°
Beilstein Reference: 1869084