L17025.03
L17025.03EA
131
EUR
InStock
L17025.03
Ethyl-3-amino-4,4,4-trifluorobutyrate 95%
Ethyl-3-amino-4,4,4-trifluorobutyrate
Purity: 95%
CAS No.: 170804-18-9
Molecular Formula: C6H10F3NO2
Formula Weight: 185.15
Refractive Index: 1.3831
Boiling Point: 70-71°/4mm
Beilstein Reference: 6136102
UN No.: UN2735
MDL No.: MFCD01075754