L17584.AE
L17584.AP
L17584.0F
L17584.AEEA
24.5
EUR
InStock
L17584.AE
L17584.AP
L17584.0F
5-Methyl-2-hexanon ≥99%
5-Methyl-2-hexanon
Purity: 99%
CAS No.: 110-12-3
Molecular Formula: C7H14O
Formula Weight: 114.19
Flash Point: 41°(105°F)
Refractive Index: 1.4075
Melting Point: -74°
Boiling Point: 144-145°
Beilstein Reference: 506163
UN No.: 2302
MDL No.: MFCD00008950