L17651.06
L17651.14
L17651.06EA
102
EUR
InStock
L17651.06
L17651.14
tert-Butyl(diethylphosphono)acetat 95%
tert-Butyl(diethylphosphono)acetat
Purity: 95%
CAS No.: 27784-76-5
Molecular Formula: C10H21O5P
Formula Weight: 252.25
Flash Point: 110°(230°F)
Refractive Index: 1.4310
Boiling Point: 100-103°/1.5mm
Beilstein Reference: 2050126
MDL No.: MFCD00075414