L17723.03
L17723.06
L17723.03EA
44.8
EUR
InStock
L17723.03
L17723.06
5-Fluor-2-iodanilin ≥97%
5-Fluor-2-iodanilin
Purity: 97%
CAS No.: 255724-71-1
Molecular Formula: C6H5FIN
Formula Weight: 237.02
Melting Point: 40-42°
Beilstein Reference: 8478850
UN No.: 2811
MDL No.: MFCD02093953