L17974.36
L17974.14
L17974.22
L17974.36EA
221
EUR
InStock
L17974.36
L17974.14
L17974.22
N,N'-Dibenzylethylendiamindiacetat 99%
N,N'-Dibenzylethylendiamindiacetat
Purity: 99%
CAS No.: 122-75-8
Molecular Formula: C6H5CH2NHCH2CH2NHCH2C6H5.2CH3CO2
Formula Weight: 360.46
Melting Point: 117-118°
Beilstein Reference: 3781362
MDL No.: MFCD00040588