L18983.06
L18983.14
L18983.06EA
74.4
EUR
InStock
L18983.06
L18983.14
o-Toluolsulfonylchlorid 80%
o-Toluolsulfonylchlorid
Purity: 97%
CAS No.: 133-59-5
Molecular Formula: C7H7ClO2S
Formula Weight: 190.65
Flash Point: 110°(230°F)
Refractive Index: 1.5560
Melting Point: 10°
Boiling Point: 154°/10mm
Beilstein Reference: 973180
UN No.: 3265
MDL No.: MFCD00024874