L19320.03
L19320.06
L19320.03EA
58.7
EUR
InStock
L19320.03
L19320.06
2'-Brom-4'-fluoracetophenon ≥98%
2'-Brom-4'-fluoracetophenon
Purity: 98%
CAS No.: 1006-39-9
Molecular Formula: C8H6BrFO
Formula Weight: 217.04
Flash Point: 110°(230°F)
Refractive Index: 1.5490
Boiling Point: 150°/12mm
Beilstein Reference: 2249797
MDL No.: MFCD00077464