L19427.03
L19427.06
L19427.03EA
37.2
EUR
InStock
L19427.03
L19427.06
2-Chinolincarbonitril ≥97%
2-Chinolincarbonitril
Purity: 97%
CAS No.: 1436-43-7
Molecular Formula: C10H6N2
Formula Weight: 154.17
Odour: Odourless
Melting Point: 93-96°
Boiling Point: 160°/23mm
Beilstein Reference: 115213
MDL No.: MFCD00134341