L19948.03
L19948.06
L19948.14
L19948.03EA
47.6
EUR
InStock
L19948.03
L19948.06
L19948.14
2-Methyl-2-propanyl-(6-aminohexyl)carbamat 98%
N-Boc-1,6-diaminohexane
Purity: 98%
CAS No.: 51857-17-1
Molecular Formula: (CH3)3COCONH(CH2)6NH2
Formula Weight: 216.33
Flash Point: 110°(230°F)
Refractive Index: 1.4620
Boiling Point: 106-110°/0.3mm
Beilstein Reference: 2089264
UN No.: UN2735
MDL No.: MFCD00671489