L19953.03
L19953.06
L19953.03EA
89
EUR
InStock
L19953.03
L19953.06
4-(Brommethyl)phenylboronsäure tech. 85%, technische Qualität
4-(Brommethyl)phenylboronsäure
Purity: 85%
CAS No.: 68162-47-0
Molecular Formula: C7H8BBrO2
Formula Weight: 214.85
Melting Point: ca 145-150°
Beilstein Reference: 3030979
UN No.: 3261
MDL No.: MFCD01318113