L20169.03
L20169.06
L20169.03EA
81.7
EUR
InStock
L20169.03
L20169.06
3-Cyanbenzolsulfonylchlorid ≥97%
3-Cyanbenzolsulfonylchlorid
Purity: 97%
CAS No.: 56542-67-7
Molecular Formula: C7H4ClNO2S
Formula Weight: 201.63
Flash Point: 110°(230°F)
Melting Point: 47-52°
Boiling Point: 114-116°/0.2mm
Beilstein Reference: 2832925
UN No.: 3261
MDL No.: MFCD00203480