L20182.06
L20182.14
L20182.06EA
73.5
EUR
InStock
L20182.06
L20182.14
(1H-Imidazol-4-yl)methanol Hydrochlorid ≥98%
(1H-Imidazol-4-yl)methanol Hydrochlorid
Purity: 98+%
CAS No.: 32673-41-9
Molecular Formula: C4H6N2O.HCl
Formula Weight: 134.57
Storage Temperature: 2°C to 8°C
Melting Point: 104-108°
Beilstein Reference: 3562041
MDL No.: MFCD00012697