L20215.03
L20215.06
L20215.14
L20215.03EA
49.9
EUR
InStock
L20215.03
L20215.06
L20215.14
Tributylvinylzinn 96%
Tributylvinylzinn
Purity: 96%
CAS No.: 7486-35-3
Molecular Formula: C14H30Sn
Formula Weight: 317.11
Flash Point: 110°(230°F)
Refractive Index: 1.4780
Boiling Point: 104-106°/3.5mm
Beilstein Reference: 3537662
UN No.: 2788
MDL No.: MFCD00009421