L20253.06
L20253.14
L20253.06EA
64.4
EUR
InStock
L20253.06
L20253.14
Samarium(III)trifluormethansulfonat 98%
Samarium(III)trifluormethansulfonat
Purity: 98%
CAS No.: 52093-28-4
Molecular Formula: Sm(SO3CF3)3
Formula Weight: 597.56
MDL No.: MFCD00192431