L20394.06
L20394.14
L20394.06EA
64.5
EUR
InStock
L20394.06
L20394.14
4-tert-Butylbenzylamin 98%
4-tert-Butylbenzylamin
Purity: 98%
CAS No.: 39895-55-1
Molecular Formula: C11H17N
Formula Weight: 163.26
Flash Point: 107°(224°F)
Refractive Index: 1.5210
Boiling Point: 124°/16mm
Beilstein Reference: 2961797
UN No.: 2735
MDL No.: MFCD00040754