M-Clarity™ Qualitätsstufe = MQ100
L6254-100G
L6254-25G
L6254-100GEA
295
EUR
InStock
L6254-100G
L6254-25G
Leishman's Eosin-Methylenblau für die Histologie, Farbstoff, Sigma-Aldrich®
Leishman's Eosin-Methylenblau
Leishman stain is a Romanowsky-type dye, similar to Wright stain. Leishman stain is used in hematology and tropical medicine to differentiate and identify leukocytes, malarial parasites, and trypanosomes. It is also known as eosin-polychrome methylene blue.
Leishman′s stain has been used for staining blood smears to differentiate blood corpuscles, malarial parasites, trypanosomers, etc. It may be used as an alternative to Giemsa. Leishman’s stain has been used for the identification of macrophage nuclei in cells.It has also been used in hematology study.