Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Microplate readers, Varioskan™ LUX

Lieferant: THERMO LABSYSTEMS LIFE SCIENCE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

VLBLATD1 VLBLATD0 VLBLATD2 VLBLA0D2 VLBLA0D1 VLBLA0D0 VLBL0TD2 VLBL0TD1 VLBL0TD0 VLBL00D2 VLBL00D1 VLBL00D0 VLB000D2 VLB000D1 VLB000D0 VL0LATD2 VL0LATD1 VL0LATD0 VL0LA0D2 VL0LA0D1 VL0LA0D0 VL0L0TD2 VL0L0TD1 VL0L0TD0 VL0L00D2 VL0L00D1 VL0000D2 VL0000D1 VL0000D0 VL0L00D0 VLBL0TGD0 VLB000GD0 VLB000GD1 VLB000GD2 VLBL00GD0 VLBL00GD1 VLBL00GD2 VLBL0TGD1 VLBL0TGD2 VLBLA0GD0 VLBLA0GD1 VLBLA0GD2 VLBLATGD0 VLBLATGD1 VLBLATGD2
735-0323EA 63200 EUR InStock
735-0323 735-0322 735-0324 735-0321 735-0320 735-0319 735-0318 735-0317 735-0316 735-0315 735-0314 735-0313 735-0312 735-0311 735-0310 735-0309 735-0308 735-0307 735-0306 735-0305 735-0304 735-0303 735-0302 735-0301 735-0300 735-0299 735-0298 735-0297 735-0296 735-0295 LABLVLBL0TGD0 LABLVLB000GD0 LABLVLB000GD1 LABLVLB000GD2 LABLVLBL00GD0 LABLVLBL00GD1 LABLVLBL00GD2 LABLVLBL0TGD1 LABLVLBL0TGD2 LABLVLBLA0GD0 LABLVLBLA0GD1 LABLVLBLA0GD2 LABLVLBLATGD0 LABLVLBLATGD1 LABLVLBLATGD2
Microplate readers, Varioskan™ LUX
Mikrotiterplatten-Photometer
Varioskan™ LUX comes equipped with a range of measurement technologies including absorbance, fluorescence intensity and FRET as standard, and with optional luminescence, AlphaScreen and time-resolved fluorescence (TRF) modules. The instrument selects the measurement wavelength using filters or monochromators, depending on which is optimal for each measurement technology.

  • Automatic dynamic range selects optimal reading range based on signal intensity
  • Smart safety controls help protect instrument and samples from user error
  • Autocalibration and self-diagnostics for confidence in results
  • Accurate temperature control for temperature critical applications
  • Integrated gas control module (optional) for setting CO₂ and O₂ concentrations for cell based assays
  • Optional dispensers to allow simultaneous dispensing and measurement for signal monitoring at the start of a reaction

SkanIt software (available in 9 languages):
Intuitive user-interface simplifies measurement setup
Virtual pipette™ tool makes it easy to define samples to plate layout
Built-in calculation options ease data processing
Manual or automatic data export to any location
Measurement data continuously saved to the database; helps prevent data loss due to interruptions such as power outage or accidental aborting
Available in 2 editions: Research Edition (included) or Drug Discovery Edition

Absorbance:
Plate types: 6 to 384 well plates
Wavelength selection: Double monochromators
Wavelength range: 200 to 1000 nm
Light source: Xenon flash lamp
Read out range: 0 to 6 Abs
Linear measurement range: 0 to 4 Abs (96-well plate) at 450 nm, ±2%; 0 to 3 Abs (384-well plate) at 450 nm, ±2%
Accuracy: 0,003 Abs or ±2%, at 200 to399 nm (0 to 2 Abs); 0,003 Abs or ±1%, at 400 to 1000 nm (0 to 3 Abs);
Precision: SD <0,001 Abs or CV <0,5%, at 450 nm (0 to 3 Abs)

Fluorescence intensity:
Plate types: 6 to 1536 well plates
Wavelength selection: Double excitation and emission monochromators
Excitation wavelength range: 200 to 1000 nm
Emission wavelength range: 270 to 840 nm
Light source: Xenon flash lamp
Sensitivity: Top reading: <0,4 fmol fluorescein/well (black 384-well plate); Bottom reading: <4 fmol fluorescein/well, (clear bottom black 384-square well plate)
Dynamic range: Top reading >6 decades; Bottom reading >5,5 decades

Time-resolved fluorescence:
Plate types: 6 to 1536 well plates
Wavelength selection: Filters (spectral scanning with double excitation and emission monochromators)
Excitation wavelength range: Fixed to 334 nm (spectral scanning 200 to 840 nm)
Emission wavelength range: 400 to 700 nm (spectral scanning 270 to 840 nm)
Light source: Xenon flash lamp
Sensitivity: <1 amol Eu/well (white low volume 384 well plate)
Dynamic range: >6 decades

Luminescence:
Plate types: 6 to 1536 well plates (spectral scanning 6 to 384 well plates)
Wavelength selection: Direct or filters (spectral scanning with double monochromators)
Wavelength range: 360 to 670 nm
Sensitivity: <7 amol ATP/well (white 384-well plate)
Dynamic range: >7 decades

AlphaScreen:
Plate types: 6 to 1536 well plates
Wavelength selection: Filters
Excitation wavelength range: Fixed to 680 nm
Emission wavelength range: 400 to 660 nm
Light source: LED
Sensitivity: <100 amol phosphotyrosine/well (white 384-well plate)

Dispensing:
Plate types: 6 to 384 well plates
No. of dispensers: None, one or two
Syringe size: 1 ml (standard), 5 ml (optional)
Dispensing volume: 2 to 5000 μl, in 1 µl increments (1 ml syringe); 5 to 25 000 μl, in 5 µl increments (5 ml syringe)
Accuracy: <1 μl with 50 μl (0,4 mm tip), <0,2 μl with 5 μl (0,25 mm tip)
Precision: <1 μl with 50 μl (0,4 mm tip), <0,25 μl with 5 μl (0,25 mm tip)
Dead volume: Reagent loss <100 μl; Total tubing volume <800 μl

Bestellinformation: For further technical specifications relating to all the available modules for the Varioskan® LUX and onboard dispenser(s), incubator and shaker please contact your local Avantor sales office.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge