Regal, Drum storage system, with water protection compartment and 1 level of shelves, with sump capacity 450 l, Inhalt: 6 - 10×200 l drums, B×T×H: 3130×1390×830 mm
Lieferant: LACONT UMWELTTECHNIK
Regal, Drum storage system, with water protection compartment and 1 level of shelves, with sump capacity 450 l, Inhalt: 6 - 10×200 l drums, B×T×H: 3130×1390×830 mm
PRODUKT
Erweitert / Kompakt
WASSERSCHUTZ-FACHCONTAINER 1 * 1 ST LACOC22-5501-B 1 * 1 ST
Hersteller-Artikelnummer:: C22-5501-B
Lokale Artikelnummer:: LACOC22-5501-B
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder