Safety storage cabinets for small gas cylinders, K-GFS
Lieferant: LACONT UMWELTTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C27-3200-B C27-3201-B
LACOC27-3200-BEA 0 EUR InStock
LACOC27-3200-B LACOC27-3201-B
Safety storage cabinets for small gas cylinders, K-GFS
Möbel Lagerschränke
Safety cabinets made of galvanised sheet steel for storing propane gas cylinders outdoors.
  • Stable floor and intermediate shelf as storage areas
  • Doors with cylinder lock to prevent unauthorised access
  • Closed back wall and roof
  • Side walls and doors with pattern of holes for stipulated ventilation
  • Marked with warning symbol
Lieferumfang: Dismounted delivery for self-assembly according to supplied instructions.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge