Sumps for small containers, PE-EP
Lieferant: LACONT UMWELTTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
P70-7083-A
LACOP70-7083-AEA 0 EUR InStock
LACOP70-7083-A
Sumps for small containers, PE-EP
Fasslager
Ideal for the storage of water-endangering and aggressive media.
  • Rotationally-moulded body of red PE
  • Circumferential stabilising ribs
  • Equipped with protective cover
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge